2,2,4,4,6,6-Hexamethylcyclotrisilazane ≥97.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
H0909-25ML
H0909-5ML
TCIAH0909-25MLEA
772
DKK
TCIAH0909-25ML
TCIAH0909-5ML
2,2,4,4,6,6-Hexamethylcyclotrisilazane ≥97.0% (by GC, titration analysis)
2,2,4,4,6,6-Hexamethylcyclotrisilazane
Formel:
C₆H₂₁N₃Si₃ Kogepunkt: 188°C Massefylde: 0,9196 g/cm³ (20 °C) Flammepunkt: 61°C |
MDL Number:
MFCD00039698 CAS-nummer: 1009-93-4 EINECS: 213-773-6 UN nummer: 1760 ADR: 8,III |
|
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Purity (Nonaqueous Titration) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...