myo-Inositol ≥99.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: i-Inositol, D-Myo-Inositol, rel-(1R,2r,3S,4R,5s,6S)-Cyclohexane-1,2,3,4,5,6-hexaol, Inositol

I0040-25G I0040-500G
TCIAI0040-25GEA 132 DKK
TCIAI0040-25G TCIAI0040-500G
myo-Inositol ≥99.0% (by GC)
myo-Inositol
Formel: C₆H₁₂O₆
Molekylvægt: 180.16 g/mol
Kogepunkt: 291
Smeltepunkt: 210
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00077932
CAS-nummer: 87-89-8
EINECS: 201-781-2
Merck Index: 14,04978
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=460e54b051b2cae75ac0c50c80d3a1aea1c74bae&vid=460e54b051b2cae75ac0c50c80d3a1aea1c74bae&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White powder to crystal
Purity (GC) min. 99.0 %
Solubility in Water almost transparency
Suitability for protein-analysis to pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR