2,4,6-Triisopropylbenzenesulfonylchlorid ≥97.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 2,4,6-Triisopropylbenzene-1-sulphonyl chloride

T0459-25G T0459-500G
TCIAT0459-25GEA 230 DKK
TCIAT0459-25G TCIAT0459-500G
2,4,6-Triisopropylbenzenesulfonylchlorid ≥97.0% (by titrimetric analysis)
2,4,6-Triisopropylbenzenesulfonylchlorid
Formel: C₁₅H₂₃ClO₂S
Smeltepunkt: 93…96 °C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00007433
CAS-nummer: 6553-96-4
EINECS: 229-479-6
UN nummer: 3261
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8c965f922dc1c18b2be82a2af42a209d682f9cfb&vid=8c965f922dc1c18b2be82a2af42a209d682f9cfb&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White powder to lump
Purity (GC) min. 95.0 %
Purity (Argentmetric Titration) min. 97.0 %
Melting point 94.0 to 99.0 deg-C
Solubility in Toluene almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR