4,4'-Dimethylbiphenyl ≥97.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
4,4'-Bitolyl, 4,4'-Dimethyldiphenyl
D1234-1G
D1234-25G
D1234-5G
TCIAD1234-1GEA
132
DKK
TCIAD1234-1G
TCIAD1234-25G
TCIAD1234-5G
4,4'-Dimethylbiphenyl ≥97.0% (by GC)
4,4'-Dimethylbiphenyl
Formel:
C₁₄H₁₄ Molekylvægt: 182.27 g/mol Kogepunkt: 295 |
MDL Number:
MFCD00008545 CAS-nummer: 613-33-2 EINECS: 210-337-7 |
|
Specifikationer Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 120.0 to 124.0 deg-C |
Solubility in Toluene | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...