3,3'-Dimethylbiphenyl 98,0 GC_ASSAY_METHOD

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 3,3'-Bitolyl, 3,3'-Dimethyl-1,1'-biphenyl

D1430-25ML D1430-5ML
TCIAD1430-25MLEA 2440 DKK
TCIAD1430-25ML TCIAD1430-5ML
3,3'-Dimethylbiphenyl 98,0 GC_ASSAY_METHOD
3,3'-Dimethylbiphenyl
Formel: C₁₄H₁₄
Molekylvægt: 182.27 g/mol
Kogepunkt: 155°C
MDL Number: MFCD00008534
CAS-nummer: 612-75-9
EINECS: 210-319-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e6acbba83b0cb66caed2944442f27804eb150043&vid=e6acbba83b0cb66caed2944442f27804eb150043&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Amber to Dark green clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR