4-(Hydroxymethyl)benzyl alkohol ≥99.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
D0605-25G
D0605-500G
D0605-100G
TCIAD0605-25GEA
189
DKK
TCIAD0605-25G
TCIAD0605-500G
TCIAD0605-100G
4-(Hydroxymethyl)benzyl alkohol ≥99.0% (by GC)
4-(Hydroxymethyl)benzyl alkohol
Formel:
C₈H₁₀O₂ Kogepunkt: 329 °C (1013 hPa) Smeltepunkt: 116…117 °C |
MDL Number:
MFCD00004665 CAS-nummer: 589-29-7 EINECS: 209-641-2 |
|
Specifikationer Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 99.0 % |
Melting point | 117.0 to 121.0 deg-C |
Solubility in hot Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...