4-Methylbenzylalkohol ≥99.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
M0162-25G
M0162-500G
M0162-100G
TCIAM0162-25GEA
132
DKK
TCIAM0162-25G
TCIAM0162-500G
TCIAM0162-100G
4-Methylbenzylalkohol ≥99.0% (by GC)
4-Methylbenzylalkohol
Formel:
C₈H₁₀O Kogepunkt: 116°C Smeltepunkt: 59 °C Massefylde: 0,978 g/cm³ (72 °C) |
MDL Number:
MFCD00004664 CAS-nummer: 589-18-4 EINECS: 209-639-1 |
|
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 99.0 % |
Melting point | 58.0 to 61.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...