4-Isopropylbenzyl alkohol ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

I0272-100ML I0272-25ML
TCIAI0272-100MLEA 870 DKK
TCIAI0272-100ML TCIAI0272-25ML
4-Isopropylbenzyl alkohol ≥98.0% (by GC)
4-Isopropylbenzyl alkohol
Formel: C₁₀H₁₄O
Kogepunkt: 249 °C (1013 hPa)
Smeltepunkt: 28 °C
Massefylde: 0,978 g/cm³ (20 °C)
MDL Number: MFCD00004663
CAS-nummer: 536-60-7
EINECS: 208-640-4
Merck Index: 14,02619
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d243b0b0263cdd853210373ed6e6b4cf91d7372e&vid=d243b0b0263cdd853210373ed6e6b4cf91d7372e&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR