4-(Trifluoromethyl)benzyl alkohol ≥96.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
(4-(Trifluoromethyl)phenyl)methanol, 4-(Hydroxymethyl)benzotrifluoride
T1312-25G
T1312-5G
TCIAT1312-25GEA
714
DKK
TCIAT1312-25G
TCIAT1312-5G
4-(Trifluoromethyl)benzyl alkohol ≥96.0% (by GC)
4-(Trifluoromethyl)benzyl alkohol
Formel:
C₈H₇F₃O Molekylvægt: 176.14 g/mol Kogepunkt: 80°C Smeltepunkt: 23 |
MDL Number:
MFCD00004661 CAS-nummer: 349-95-1 EINECS: 206-494-6 |
|
Specifikationer Testresultater
Appearance | White or Colorless to Light yellow powder to lump to clear liquid |
Purity (GC) | min. 96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...