3-Hydroxybenzylalkohol ≥99.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
H0918-25G
H0918-5G
TCIAH0918-25GEA
1060
DKK
TCIAH0918-25G
TCIAH0918-5G
3-Hydroxybenzylalkohol ≥99.0% (by GC)
3-Hydroxybenzylalkohol
Formel:
C₇H₈O₂ Kogepunkt: 123°C Smeltepunkt: 70 °C Massefylde: 1,161 g/cm³ (25 °C) |
MDL Number:
MFCD00004643 CAS-nummer: 620-24-6 EINECS: 210-633-6 |
|
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 99.0 % |
Melting point | 70.0 to 73.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...