3,4,5-Trimethoxybenzylalkohol ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 2,4,6-Trimethoxybenzyl alcohol, (3,4,5-Trimethoxyphenyl)methanol

T1103-25G T1103-500G
TCIAT1103-25GEA 435 DKK
TCIAT1103-25G TCIAT1103-500G
3,4,5-Trimethoxybenzylalkohol ≥98.0% (by GC)
3,4,5-Trimethoxybenzylalkohol
Formel: C₁₀H₁₄O₄
Kogepunkt: 155°C
Smeltepunkt: 36…38 °C
Massefylde: 1,1427 g/cm³ (20 °C)
MDL Number: MFCD00060356
CAS-nummer: 3840-31-1
EINECS: 223-331-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f295ced08085a66ca19d3faa5d14b89c3500dbfb&vid=f295ced08085a66ca19d3faa5d14b89c3500dbfb&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR