1-tert-Butyl-4-iodbenzen ≥95.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

B3624-25G B3624-5G
TCIAB3624-25GEA 2040 DKK
TCIAB3624-25G TCIAB3624-5G
1-tert-Butyl-4-iodbenzen ≥95.0% (by GC)
1-tert-Butyl-4-iodbenzen
Formel: C₁₀H₁₃I
Kogepunkt: 118°C
Massefylde: 1,4392 g/cm³ (14 °C)
MDL Number: MFCD00052339
CAS-nummer: 35779-04-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ce815e98c86261adae892e8b7ec82181027f7827&vid=ce815e98c86261adae892e8b7ec82181027f7827&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR