3-(Trifluormethyl)benzensulfonylchlorid ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
3-(Trifluoromethyl)benzene-1-sulphonyl chloride
T1910-25G
T1910-5G
TCIAT1910-25GEA
1200
DKK
TCIAT1910-25G
TCIAT1910-5G
3-(Trifluormethyl)benzensulfonylchlorid ≥98.0% (by GC, titration analysis)
3-(Trifluormethyl)benzensulfonylchlorid
Formel:
C₇H₄ClF₃O₂S Molekylvægt: 244.62 g/mol |
MDL Number:
MFCD00014724 CAS-nummer: 777-44-6 UN nummer: 3265 ADR: 8 III |
|
Specifikationer Testresultater
Appearance | White to Yellow to Green clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...