4-(Trifluormethyl)anthranilsyre ≥97.0% (by HPLC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
A2175-25G
A2175-5G
TCIAA2175-25GEA
1900
DKK
TCIAA2175-25G
TCIAA2175-5G
4-(Trifluormethyl)anthranilsyre ≥97.0% (by HPLC, titration analysis)
4-(Trifluormethyl)anthranilsyre
Formel:
C₈H₆F₃NO₂ Smeltepunkt: 175°C |
MDL Number:
MFCD00519334 CAS-nummer: 402-13-1 EINECS: 206-937-3 |
|
Specifikationer Testresultater
Appearance | White to Amber powder to crystalline |
Purity (HPLC) | min. 97.0 area% |
Purity (Neutralization titration) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...