(4-Fluorphenyl)acetonitril ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 4-Fluorobenzyl cyanide, 4-Fluorophenylacetonitrile

F0258-25G F0258-250G
TCIAF0258-25GEA 255 DKK
TCIAF0258-25G TCIAF0258-250G
(4-Fluorphenyl)acetonitril ≥98.0% (by GC)
(4-Fluorphenyl)acetonitril
Formel: C₈H₆FN
Molekylvægt: 135.14 g/mol
Kogepunkt: 230°C
Smeltepunkt: 86
Massefylde: 1.136
Flammepunkt: 108°C
MDL Number: MFCD00001917
CAS-nummer: 459-22-3
EINECS: 207-286-8
UN nummer: 3276
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6cf22c7328ce46acf996637aab431853a7b0fead&vid=6cf22c7328ce46acf996637aab431853a7b0fead&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR