4-Methyl-m-benzoesyre ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 3-Methoxy-4-methylbenzoic acid

M1426-25G M1426-5G
TCIAM1426-25GEA 1950 DKK
TCIAM1426-25G TCIAM1426-5G
4-Methyl-m-benzoesyre ≥98.0% (by HPLC, titration analysis)
4-Methyl-m-benzoesyre
Formel: C₉H₁₀O₃
Molekylvægt: 166.18 g/mol
MDL Number: MFCD00002566
CAS-nummer: 7151-68-0
EINECS: 230-486-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f112dcedb33dca25a587a2ca1509e84c14be5ecc&vid=f112dcedb33dca25a587a2ca1509e84c14be5ecc&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Solubility in Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR