2-Methyl-m-benzoesyre ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 3-Methoxy-2-methylbenzoic acid

M2217-25G M2217-5G
TCIAM2217-25GEA 427 DKK
TCIAM2217-25G TCIAM2217-5G
2-Methyl-m-benzoesyre ≥98.0% (by HPLC, titration analysis)
2-Methyl-m-benzoesyre
Formel: C₉H₁₀O₃
Molekylvægt: 166.18 g/mol
MDL Number: MFCD02094039
CAS-nummer: 55289-06-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c896ae6565a786a0e6677f9b435891cc1866f89d&vid=c896ae6565a786a0e6677f9b435891cc1866f89d&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Yellow to Orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 147.0 to 151.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR