ω,ω-Diethoxyacetophenon ≥95.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 2,2-Diethoxyacetophenone, 2,2-Diethoxy-1-phenylethan-1-one

D1640-25G D1640-500G
TCIAD1640-25GEA 214 DKK
TCIAD1640-25G TCIAD1640-500G
ω,ω-Diethoxyacetophenon ≥95.0% (by GC)
ω,ω-Diethoxyacetophenon
Formel: C₁₂H₁₆O₃
Molekylvægt: 208.26 g/mol
Kogepunkt: 134°C
Flammepunkt: 127°C
MDL Number: MFCD00009659
CAS-nummer: 6175-45-7
EINECS: 228-220-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fca07ed4947ece4afa36da1809151ffab79df8fb&vid=fca07ed4947ece4afa36da1809151ffab79df8fb&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Red to Green clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR