(4-Chlorphenyl)acetonitril ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 4-Chlorobenzyl cyanide

C0585-25G C0585-500G
TCIAC0585-25GEA 132 DKK
TCIAC0585-25G TCIAC0585-500G
(4-Chlorphenyl)acetonitril ≥98.0% (by GC)
(4-Chlorphenyl)acetonitril
Formel: C₈H₆ClN
Kogepunkt: 267°C
Massefylde: 1,1968 g/cm³ (20 °C)
Flammepunkt: 133 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00001918
CAS-nummer: 140-53-4
EINECS: 205-418-9
UN nummer: 3439
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4c915b4925d872744420c4c8d7559b33d96acf65&vid=4c915b4925d872744420c4c8d7559b33d96acf65&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR