α-Brom-4-Chlor-2-Fluortoluen ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
4-Chloro-2-fluorobenzyl bromide, 1-(Bromomethyl)-4-chloro-2-fluorobenzene
C2483-25G
C2483-5G
TCIAC2483-25GEA
1560
DKK
TCIAC2483-25G
TCIAC2483-5G
α-Brom-4-Chlor-2-Fluortoluen ≥98.0% (by GC, titration analysis)
α-Brom-4-Chlor-2-Fluortoluen
Formel:
C₇H₅BrClF Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD00831105 CAS-nummer: 71916-82-0 UN nummer: 3265 ADR: 8,II |
|
Specifikationer Testresultater
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...