α-Brom-2-Chlor-5-Fluortoluen ≥96.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

C2359-1G C2359-5G
TCIAC2359-1GEA 222 DKK
TCIAC2359-1G TCIAC2359-5G
α-Brom-2-Chlor-5-Fluortoluen ≥96.0% (by GC)
α-Brom-2-Chlor-5-Fluortoluen
Formel: C₇H₅BrClF
Kogepunkt: 66°C
Flammepunkt: 18°C
MDL Number: MFCD01631419
CAS-nummer: 81778-09-8
UN nummer: 2924
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2267f11d57aeb0a41c65ad48274b53b210375547&vid=2267f11d57aeb0a41c65ad48274b53b210375547&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR