1-Brom-4-octylbenzen ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
(4-Bromophenyl)octane
B2117-5G
B2117-25G
TCIAB2117-5GEA
764
DKK
TCIAB2117-5G
TCIAB2117-25G
1-Brom-4-octylbenzen ≥98.0% (by GC)
1-Brom-4-octylbenzen
Formel:
C₁₄H₂₁Br Kogepunkt: 125°C |
MDL Number:
MFCD00038348 CAS-nummer: 51554-93-9 |
|
Specifikationer Testresultater
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...