1-Brom-4-ethynylbenzen ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 4-Bromophenylacetylene, 1-Ethynyl-4-bromobenzene, (4-Bromophenyl)acetylene, (4-Bromophenyl)ethyne

B3701-1G B3701-5G
TCIAB3701-1GEA 238 DKK
TCIAB3701-1G TCIAB3701-5G
1-Brom-4-ethynylbenzen ≥98.0% (by GC)
1-Brom-4-ethynylbenzen
Formel: C₈H₅Br
Kogepunkt: 92°C
MDL Number: MFCD00168822
CAS-nummer: 766-96-1
UN nummer: 3077
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=103a5795789c4e18548491316488b0345e70e5a7&vid=103a5795789c4e18548491316488b0345e70e5a7&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 64.0 to 68.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR