2-Nitro-p-phenylendiamin ≥94.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-Nitro-1,4-phenylenediamine, 2-Nitrobenzene-1,4-diamine

D0105-500G D0105-25G
TCIAD0105-500GEA 1470 DKK
TCIAD0105-500G TCIAD0105-25G
2-Nitro-p-phenylendiamin ≥94.0% (by titrimetric analysis)
2-Nitro-p-phenylendiamin
Formel: C₆H₇N₃O₂
Smeltepunkt: 134…137 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00007903
CAS-nummer: 5307-14-2
EINECS: 226-164-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2b89134db7531aec8b10f3461bdfdf47e417da4a&vid=2b89134db7531aec8b10f3461bdfdf47e417da4a&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Dark green to Dark blue to Black powder to crystal
Purity (Nonaqueous Titration) min. 94.0 %
Melting point 136.0 to 141.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR