Methyl2-iodbenzoat ≥98.0% (by GC)
Leverandør: TCI
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
Synonymer: Methyl 2-iodobenzoate, 2-Iodobenzoic acid methyl ester
I0586-25G I0586-250G
TCIAI0586-25GEA 247 DKK
TCIAI0586-25G TCIAI0586-250G
Methyl2-iodbenzoat ≥98.0% (by GC)
Methyl2-iodbenzoat
Formel: C₈H₇IO₂
Molekylvægt: 262.05 g/mol
Kogepunkt: 274°C
MDL Number: MFCD00016351
CAS-nummer: 610-97-9
EINECS: 210-243-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=21f861f07991784ed1a79b9cc3240b42e9e93000&vid=21f861f07991784ed1a79b9cc3240b42e9e93000&prid=web&clid=SAPDAM
Bestil nu
Specifikationer Testresultater
Appearance Light orange to Yellow to Green clear liquid
Purity (GC) min. 98.0 %