Pentafluorbenzensulfonylchlorid ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
2,3,4,5,6-Pentafluorobenzenesulphonyl chloride, Pentafluorobenzene-1-sulphonyl chloride
P0934-25G
P0934-5G
TCIAP0934-25GEA
1150
DKK
TCIAP0934-25G
TCIAP0934-5G
Pentafluorbenzensulfonylchlorid ≥98.0% (by GC)
Pentafluorbenzensulfonylchlorid
Formel:
C₆ClF₅O₂S Kogepunkt: 210°C Massefylde: 1,79 g/cm³ (20 °C) Flammepunkt: 90°C |
MDL Number:
MFCD00007427 CAS-nummer: 832-53-1 EINECS: 212-620-0 UN nummer: 3265 ADR: 8,II |
|
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...