3-(Trifluormethoxy)phenol ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
α,α,α-Trifluoro-3-hydroxyanisole
T1615-25G
T1615-5G
TCIAT1615-25GEA
1390
DKK
TCIAT1615-25G
TCIAT1615-5G
3-(Trifluormethoxy)phenol ≥98.0% (by GC)
3-(Trifluormethoxy)phenol
Formel:
C₇H₅F₃O₂ Molekylvægt: 178.11 g/mol Kogepunkt: 80°C Flammepunkt: 84°C |
MDL Number:
MFCD00040987 CAS-nummer: 827-99-6 |
|
Specifikationer Testresultater
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...