1-Nitro-3-(trifluormethoxy)benzen ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
N0884-25G
N0884-5G
TCIAN0884-25GEA
682
DKK
TCIAN0884-25G
TCIAN0884-5G
1-Nitro-3-(trifluormethoxy)benzen ≥98.0% (by GC)
1-Nitro-3-(trifluormethoxy)benzen
Formel:
C₇H₄F₃NO₃ Kogepunkt: 96°C |
MDL Number:
MFCD00041011 CAS-nummer: 2995-45-1 |
|
Specifikationer Testresultater
Appearance | Light yellow to Yellow to Orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...