5-fluorsalicylsyre ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 5-Fluoro-2-hydroxybenzoic acid

F0469-1G F0469-5G
TCIAF0469-1GEA 345 DKK
TCIAF0469-1G TCIAF0469-5G
5-fluorsalicylsyre ≥98.0% (by GC, titration analysis)
5-fluorsalicylsyre
Formel: C₇H₅FO₃
Kogepunkt: 295 °C (1013 hPa)
Smeltepunkt: 180 °C
MDL Number: MFCD00002456
CAS-nummer: 345-16-4
EINECS: 206-455-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aefc457e1e034409e78050f1c4ba36af079b6c02&vid=aefc457e1e034409e78050f1c4ba36af079b6c02&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 179.0 to 183.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR