4-Fluor-3-methoxybenzoesyre ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 3-Methoxy-4-fluorobenzoic acid

F0681-1G F0681-5G
TCIAF0681-1GEA 329 DKK
TCIAF0681-1G TCIAF0681-5G
4-Fluor-3-methoxybenzoesyre ≥98.0% (by GC, titration analysis)
4-Fluor-3-methoxybenzoesyre
Formel: C₈H₇FO₃
Molekylvægt: 170.14 g/mol
MDL Number: MFCD00272134
CAS-nummer: 82846-18-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=48287dc194d6bc3829a3405c7c842961392ac74e&vid=48287dc194d6bc3829a3405c7c842961392ac74e&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 205.0 to 209.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR