2-Fluor-4-methoxybenzoesyre ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

F0572-25G F0572-5G
TCIAF0572-25GEA 1560 DKK
TCIAF0572-25G TCIAF0572-5G
2-Fluor-4-methoxybenzoesyre ≥98.0% (by titrimetric analysis)
2-Fluor-4-methoxybenzoesyre
Formel: C₈H₇FO₃
MDL Number: MFCD00509373
CAS-nummer: 394-42-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=78418d5892ed60e0da88bec5d173374d87c10f2b&vid=78418d5892ed60e0da88bec5d173374d87c10f2b&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 191.0 to 195.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR