Methyl4-nitrobenzoat ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

N0159-25G N0159-500G
TCIAN0159-25GEA 542 DKK
TCIAN0159-25G TCIAN0159-500G
Methyl4-nitrobenzoat ≥98.0% (by GC)
Methyl4-nitrobenzoat
Formel: C₈H₇NO₄
Smeltepunkt: 94…96 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00007350
CAS-nummer: 619-50-1
EINECS: 210-599-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1937263ec912e45eff349a749d65b06b05cee5b1&vid=1937263ec912e45eff349a749d65b06b05cee5b1&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 95.0 to 97.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR