Methyl3-hydroxybenzoat ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Methyl 3-Hydroxybenzoate, 3-Hydroxybenzoic acid methyl ester
H0215-25G
H0215-500G
H0215-100G
TCIAH0215-25GEA
214
DKK
TCIAH0215-25G
TCIAH0215-500G
TCIAH0215-100G
Methyl3-hydroxybenzoat ≥98.0% (by GC, titration analysis)
Methyl3-hydroxybenzoat
Formel:
C₈H₈O₃ Molekylvægt: 152.15 g/mol Kogepunkt: 178°C |
MDL Number:
MFCD00002295 CAS-nummer: 19438-10-9 EINECS: 243-071-5 |
|
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 71.0 to 74.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...