Ethyl 4-(dimethylamino)benzoat ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 4-(Dimethylamino)benzoic acid ethyl ester, Ethyl 4-dimethylaminobenzoate

D1744-500G D1744-25G D1744-100G
TCIAD1744-500GEA 1280 DKK
TCIAD1744-500G TCIAD1744-25G TCIAD1744-100G
Ethyl 4-(dimethylamino)benzoat ≥98.0% (by GC)
Ethyl 4-(dimethylamino)benzoat
Formel: C₁₁H₁₅NO₂
Molekylvægt: 193.25 g/mol
Kogepunkt: 112
MDL Number: MFCD00009115
CAS-nummer: 10287-53-3
EINECS: 233-634-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=23ecd96d9ef54f45aae1f046a1bb76e6985e39ce&vid=23ecd96d9ef54f45aae1f046a1bb76e6985e39ce&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 62.0 to 67.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR