3-Chlor-4-fluorbenzoylchlorid ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

C2122-25G C2122-5G
TCIAC2122-25GEA 2750 DKK
TCIAC2122-25G TCIAC2122-5G
3-Chlor-4-fluorbenzoylchlorid ≥98.0% (by GC, titration analysis)
3-Chlor-4-fluorbenzoylchlorid
Formel: C₇H₃Cl₂FO
Kogepunkt: 222 °C (1013 hPa)
MDL Number: MFCD00236215
CAS-nummer: 65055-17-6
UN nummer: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3c583f30efeab92fe6822c7b51d4214eb253fbea&vid=3c583f30efeab92fe6822c7b51d4214eb253fbea&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR