2-Chlor-6-fluorbenzoylchlorid ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

C2229-25G C2229-5G
TCIAC2229-25GEA 706 DKK
TCIAC2229-25G TCIAC2229-5G
2-Chlor-6-fluorbenzoylchlorid ≥98.0% (by GC, titration analysis)
2-Chlor-6-fluorbenzoylchlorid
Formel: C₇H₃Cl₂FO
Kogepunkt: 227 °C (1013 hPa)
Smeltepunkt: 57 °C
MDL Number: MFCD00178746
CAS-nummer: 79455-63-3
EINECS: 279-162-1
UN nummer: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ac7aaec4d7661e9ddc0575401a47b00e12a95177&vid=ac7aaec4d7661e9ddc0575401a47b00e12a95177&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR