4-Chlor-2-methoxybenzoesyre ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
4-Chloro-o-anisic acid
C2076-25G
C2076-5G
TCIAC2076-25GEA
911
DKK
TCIAC2076-25G
TCIAC2076-5G
4-Chlor-2-methoxybenzoesyre ≥98.0% (by GC, titration analysis)
4-Chlor-2-methoxybenzoesyre
Formel:
C₈H₇ClO₃ Kogepunkt: 308 °C (1013 hPa) Smeltepunkt: 148 °C |
MDL Number:
MFCD00002532 CAS-nummer: 57479-70-6 EINECS: 260-761-1 |
|
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Solubility in Methanol | almost transparency |
Melting point | 146.0 and le 150.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...