4-Amino-5-chlor-2-methoxybenzoesyre ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Metoclopramide Impurity C, 4-Amino-5-chloro-o-anisic acid
A1237-25G
A1237-500G
TCIAA1237-25GEA
731
DKK
TCIAA1237-25G
TCIAA1237-500G
4-Amino-5-chlor-2-methoxybenzoesyre ≥98.0% (by HPLC, titration analysis)
4-Amino-5-chlor-2-methoxybenzoesyre
Formel:
C₈H₈ClNO₃ Molekylvægt: 201.61 g/mol Kogepunkt: 354 |
MDL Number:
MFCD00038794 CAS-nummer: 7206-70-4 EINECS: 230-582-3 |
|
Specifikationer Testresultater
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...