Ethyl 3-Brombenzoat ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
Ethyl 3-bromobenzoate, 3-Bromobenzoic acid ethyl ester
B1808-25G
B1808-5G
TCIAB1808-25GEA
608
DKK
TCIAB1808-25G
TCIAB1808-5G
Ethyl 3-Brombenzoat ≥98.0% (by GC)
Ethyl 3-Brombenzoat
Formel:
C₉H₉BrO₂ Molekylvægt: 229.07 g/mol Kogepunkt: 261°C |
MDL Number:
MFCD00013529 CAS-nummer: 24398-88-7 EINECS: 246-223-9 |
|
Specifikationer Testresultater
Appearance | Colorless to Red to Green clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...