2-Methylpiperidin ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
P0443-25ML
P0443-500ML
P0443-100ML
TCIAP0443-25MLEA
132
DKK
TCIAP0443-25ML
TCIAP0443-500ML
TCIAP0443-100ML
2-Methylpiperidin ≥98.0% (by GC)
2-Methylpiperidin
Formel:
C₆H₁₃N Kogepunkt: 118°C Smeltepunkt: –4,6 °C Massefylde: 0,842 g/cm³ (20 °C) Flammepunkt: 10°C |
MDL Number:
MFCD00005982 CAS-nummer: 109-05-7 EINECS: 203-642-1 UN nummer: 1993 ADR: 3,II |
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...