4-Nitrophenyl chlorformat ≥98.0% (by titrimetric analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
C1400-25G
C1400-250G
TCIAC1400-25GEA
509
DKK
TCIAC1400-25G
TCIAC1400-250G
4-Nitrophenyl chlorformat ≥98.0% (by titrimetric analysis)
4-Nitrophenyl chlorformat
Formel:
C₇H₄ClNO₄ Molekylvægt: 201.57 g/mol Kogepunkt: 162°C Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD00007321 CAS-nummer: 7693-46-1 EINECS: 231-706-9 UN nummer: 2923 ADR: 8 II |
|
Specifikationer Testresultater
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 96.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Melting point | 80.0 to 83.0 deg-C |
Solubility in Acetone | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...