Pivalinsyre ≥99.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 2,2-Dimethylpropionic acid, Trimethylacetic acid

P0461-25G P0461-400G P0461-100G
TCIAP0461-25GEA 132 DKK
TCIAP0461-25G TCIAP0461-400G TCIAP0461-100G
Pivalinsyre ≥99.0% (by GC)
Pivalinsyre
Formel: C₅H₁₀O₂
Kogepunkt: 164°C
Smeltepunkt: 32…35 °C
Massefylde: 0,91 g/cm³ (20 °C)
Flammepunkt: 640 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00004194
CAS-nummer: 75-98-9
EINECS: 200-922-5
UN nummer: 3261
ADR: 8,II
Merck Index: 14,07511
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11b4ebfd064035db1129782e4e709826f039a82a&vid=11b4ebfd064035db1129782e4e709826f039a82a&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White or Colorless to Almost white or Almost colorless powder to lump to clear liquid
Purity (GC) min. 99.0 %
Purity (Neutralization titration) min. 98.0 %
Freezing point 32.0 to 36.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR