Methansulfonsyre natriumsalt ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Methanesulphonic acid sodium salt

S0518-25G S0518-500G
TCIAS0518-25GEA 247 DKK
TCIAS0518-25G TCIAS0518-500G
Methansulfonsyre natriumsalt ≥98.0% (by titrimetric analysis)
Methansulfonsyre natriumsalt
Formel: CH₃NaO₃S
MDL Number: MFCD00064389
CAS-nummer: 2386-57-4
EINECS: 219-203-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2383cc654c8e884067f1d5998cf17055e3e6612e&vid=2383cc654c8e884067f1d5998cf17055e3e6612e&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (Ion exchange titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR