1,12-Dodecandiol ≥99.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
1,12-Dihydroxy dodecane
D0969-25G
D0969-500G
D0969-100G
TCIAD0969-25GEA
181
DKK
TCIAD0969-25G
TCIAD0969-500G
TCIAD0969-100G
1,12-Dodecandiol ≥99.0% (by GC)
1,12-Dodecanediol
Formel:
C₁₂H₂₆O₂ Kogepunkt: 189°C Smeltepunkt: 78…79 °C Massefylde: 1,08 g/cm³ (20 °C) |
MDL Number:
MFCD00004755 CAS-nummer: 5675-51-4 EINECS: 227-133-9 |
|
Specifikationer Testresultater
Appearance | White to Almost white powder to lump |
Purity (GC) | min. 99.0 % |
Melting point | 80.0 to 83.0 deg-C |
Solubility in hot Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...