2-(2-Aminoethoxy)ethanol ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
A0301-25ML
A0301-500ML
TCIAA0301-25MLEA
140
DKK
TCIAA0301-25ML
TCIAA0301-500ML
2-(2-Aminoethoxy)ethanol ≥98.0% (by GC, titration analysis)
2-(2-Aminoethoxy)ethanol
Formel:
C₄H₁₁NO₂ Molekylvægt: 105.14 g/mol Kogepunkt: 223°C Smeltepunkt: -11 Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00008181 CAS-nummer: 929-06-6 EINECS: 213-195-4 UN nummer: 3055 ADR: 8 III |
|
Specifikationer Testresultater
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...