Iodacetonitril ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

I0419-25G I0419-5G
TCIAI0419-25GEA 3970 DKK
TCIAI0419-25G TCIAI0419-5G
Iodacetonitril ≥98.0% (by GC)
Iodacetonitril
Formel: C₂H₂IN
Kogepunkt: 78°C
Massefylde: 2,3065 g/cm³ (20 °C)
Flammepunkt: 86°C
MDL Number: MFCD00001886
CAS-nummer: 624-75-9
EINECS: 210-860-0
UN nummer: 1760
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2321770da49126d379a01a2f3d094a42d0459bb0&vid=2321770da49126d379a01a2f3d094a42d0459bb0&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Yellow to Amber to Dark purple clear liquid to cloudy liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR