Ethyl trifluoracetat ≥99.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Trifluoroacetic ethyl ester

T0432-25G T0432-500G T0432-100G
TCIAT0432-25GEA 132 DKK
TCIAT0432-25G TCIAT0432-500G TCIAT0432-100G
Ethyl trifluoracetat ≥99.0% (by GC)
Ethyl trifluoracetat
Formel: C₄H₅F₃O₂
Kogepunkt: 62°C
Smeltepunkt: –78 °C
Massefylde: 1,19 g/cm³ (20 °C)
Flammepunkt: -7°C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00000419
CAS-nummer: 383-63-1
EINECS: 206-851-6
UN nummer: 2924
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=488558da2b6ced993030ce81714bf616df6e5e78&vid=488558da2b6ced993030ce81714bf616df6e5e78&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR