L-Methionin ≥99.0% (by HPLC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
(S)-(-)-2-Amino-4-(methylthio)butyric acid, (S)-(-)-Methionine, L(-)-Methionine, (S)-(-)-2-Amino-4-(methylthio)butanoic acid, H-Met-OH, Methionine, (S)-2-Amino-4-(methylthio)butanoic acid
M0099-25G
M0099-500G
M0099-100G
TCIAM0099-25GEA
148
DKK
TCIAM0099-25G
TCIAM0099-500G
TCIAM0099-100G
L-Methionin ≥99.0% (by HPLC, titration analysis)
L-Methionin
Formel:
C₅H₁₁NO₂S Kogepunkt: 278 °C (1013 hPa) Smeltepunkt: 136 °C Massefylde: 1,373 g/cm³ (20 °C) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00063097 CAS-nummer: 63-68-3 EINECS: 200-562-9 Merck Index: 14,05975 |
|
Specifikationer Testresultater
Appearance | White powder to crystal |
Purity (HPLC) | min. 99.0 area% |
Purity (Nonaqueous Titration) | min. 99.0 % |
Specific rotation [a]20/D | +23.7 to +24.7 deg (C=8, HCl (1+1)) (after drying) |
Solubility in Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...