Acetoacetanilid ≥99.0% (by HPLC, total nitrogen)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
3-Oxo-N-phenyl butamide, 3-Oxo-N-phenyl butanamide
A0046-25G
A0046-500G
TCIAA0046-25GEA
132
DKK
TCIAA0046-25G
TCIAA0046-500G
Acetoacetanilid ≥99.0% (by HPLC, total nitrogen)
Acetoacetanilid
Formel:
C₁₀H₁₁NO₂ Kogepunkt: 129 °C (32 -rr) Smeltepunkt: 130…132 °C Massefylde: 1,26 g/cm³ (20 °C) |
MDL Number:
MFCD00008780 CAS-nummer: 102-01-2 EINECS: 202-996-4 Merck Index: 14,00058 |
|
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 99.0 area% |
Purity (with Total Nitrogen) | min. 99.0 % |
Melting point | 83.0 to 86.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...