Ravsyredinitril ≥99.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
S0109-25G
S0109-500G
S0109-100G
TCIAS0109-25GEA
156
DKK
TCIAS0109-25G
TCIAS0109-500G
TCIAS0109-100G
Ravsyredinitril ≥99.0% (by GC)
Ravsyredinitril
Formel:
C₄H₄N₂ Kogepunkt: 159°C Smeltepunkt: 53 °C Massefylde: 1,023 g/cm³ (20 °C) Flammepunkt: 132 °C Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD00001949 CAS-nummer: 110-61-2 EINECS: 203-783-9 UN nummer: 3439 ADR: 6.1,III Merck Index: 14,08872 |
|
Specifikationer Testresultater
Appearance | White to Almost white powder to lump |
Purity (GC) | min. 99.0 % |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...